Objektglas med fordybning
Microscope slides with reaction wells (numbered clockwise), blå, 10, 7 mm
Katalog # MARI1216821
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Documents
Specifikationer
- Ant. brønde:10
- Farve:blå
- Beskrivelse:Microscope slides with reaction wells (numbered clockwise)
- Seq:24
- Brønd Ø:7 mm