Contrast clips
PP, 13,8 - 17,0, 36 mm, hvid
Katalog # WITG3.691.004
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
PP, white/black.
- For attachment to all measuring flasks
- With grip deepening
- Unbreakable, resistant to acids and alkalis
- For better recognition and adjustment of the liquid level
Specifikationer
- Spændvidde:13,8 - 17,0
- Farve:hvid
- Længde:36 mm
- Materiale:PP
- Seq:04