Orders LinkContactUsLinkComponent
Separating Funnels, Squibb
Separating Funnels, Squibb
50 ml, 19/26 NS
Katalog # KAVA632426355056
Leverandør:   Kavalier
Separating Funnels, Squibb
Katalog # KAVA632426355056
Leverandør:  Kavalier
Separating Funnels, Squibb
Separating Funnels, Squibb
50 ml, 19/26 NS
Price
1.500,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
Specifikationer
  • Kappestr.:
    19/26 NS
  • Kapacitet:
    50 ml
  • Seq:
    01