Order Entry
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

Tube bend, with cap nut, 19,05 mm, nozzle-Ø 12 mm, PP, 19,05 mm
Katalog # BURK8548-3412
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8548-3412
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Tube bend, with cap nut, 19,05 mm, nozzle-Ø 12 mm, PP, 19,05 mm

Price
59,90 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    19,05 mm
  • Beskrivelse:
    Tube bend, with cap nut, 19,05 mm, nozzle-Ø 12 mm
  • Materiale:
    PP
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.