Orders LinkContactUsLinkComponent
Glass Wool
Glass wool
  ACRO386060500
Glass Wool
  ACRO386060500
Glass Wool
Glass Wool
Glass wool
218,00 kr
1 * 50 g
This glass wool can be used as a filter aid.
Boiling point: >1000 °C (760 mmHg)
  • Beskrivelse:
    Glass wool