Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Gentamicin sulfat

Gentamicin sulfat

Glass bottle for solids, 10 g
Catalog # ICNA0219005710
Health
Supplier:   MP Biomedicals
Gentamicin sulfat
Catalog # ICNA0219005710
Supplier:  MP Biomedicals
CAS Number:  1405-41-0
Gentamicin sulfat

Glass bottle for solids, 10 g

CAS Number:  1405-41-0
Price
4.570,00 kr
Log in for availability
1 * 10 g
Product Details & Documents

Gentamicin sulpahte is used as a selection agent (gentamicin-resistance gene) in molecular biology applications.It is used to treat many types of bacterial infections, particularly those caused by gram-negative organisms.

It causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site. The bactericidal effect of gentamicin on Pseudomonas aeruginosa is exerted by the binding of gentamicin to the outer membrane, where it displaces natural cations, destabilises the membrane, and forms holes in the cell surface. Antimicrobial spectrum: Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria.

H2O: soluble 50 mg/ml (As a stock solution. Stock solutions should be stored at −20 °C.) (Lit.)

Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Merck Index:13,04403
  • Melting Pt:218…237 °C
  • Storage Temperature:Stue temperatur
  • EINECS:215-778-9
Specifications

Specification Test Results

  • Synonyms:
    Garamycin; Gentamicin sulphate salt; Gentiomycin C
  • Form:
    White powder
  • Storage:
    Store at +4 °C
  • Loss on Drying:
    ≤18,0 %
  • Optical Rotation:
    107° to +121° (10 mg/ml, water)
  • pH:
    3,5 - 5,5 (1 in 25, in water)
  • Potency:
    ≥590 µg/mg

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifications