Olie rød O
Glass bottle for solids, 1 kg
CAS No.: 1320-06-5 Molecular Formula: C26H24N4O Formula Weight: 408.51 Melting Point: ca 166° dec. Beilstein Reference: 3491382 MDL No.: MFCD00003898
: CI 26125
- :1320-06-5
- :C₂₆H₂₄N₄O
- :MFCD00003898
- :408.51 g/mol
- :∼ 166 °C (dec.)
- :Stue temperatur
- :215-295-3
- Pakning:Glass bottle for solids
- Pakn:1 kg