Orders LinkContactUsLinkComponent
Olie rød O
Glass bottle for solids, 1 kg
  A12989.A1
Olie rød O
  A12989.A1
 :  1320-06-5
Olie rød O
Olie rød O
Glass bottle for solids, 1 kg
 :  1320-06-5
1 * 1 kg
CAS No.: 1320-06-5 Molecular Formula: C26H24N4O Formula Weight: 408.51 Melting Point: ca 166° dec. Beilstein Reference: 3491382 MDL No.: MFCD00003898
 :  CI 26125
  •  :1320-06-5
  •  :C₂₆H₂₄N₄O
  •  :MFCD00003898
  •  :408.51 g/mol
  •  :∼ 166 °C (dec.)
  •  :Stue temperatur
  •  :215-295-3
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg