Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® Bulb Pipettes, Two Marks, Class AS

VWR® Bulb Pipettes, Two Marks, Class AS

3 ml, 0,010 ± ml, 350 mm
Katalog # 612-4777
Leverandør:   VWR Collection
VWR® Bulb Pipettes, Two Marks, Class AS
Katalog # 612-4777
Leverandør:  VWR Collection
VWR® Bulb Pipettes, Two Marks, Class AS
VWR® Bulb Pipettes, Two Marks, Class AS

3 ml, 0,010 ± ml, 350 mm

Price
180,00 kr
Log ind for tilgængelighed
1 * 3 Items
Product Details & Documents

Borosilicate 9.1 glass.

  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in high contrast amber stain

Certifications: DIN EN ISO 648

Bestillingsoplysninger: Dated batch identification and batch certificate.

Specifikationer
  • Kapacitet:
    3 ml
  • Længde:
    350 mm
  • Seq:
    03
  • Tolerance:
    0,010 ± ml