Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® Bulb Pipettes, Two Marks, Class AS

VWR® Bulb Pipettes, Two Marks, Class AS

2 ml, 0,010 ± ml, 330 mm
Catalog # 612-4776
Supplier:   VWR Collection
VWR® Bulb Pipettes, Two Marks, Class AS
Catalog # 612-4776
Supplier:  VWR Collection
VWR® Bulb Pipettes, Two Marks, Class AS
VWR® Bulb Pipettes, Two Marks, Class AS

2 ml, 0,010 ± ml, 330 mm

Price
168,00 kr
Log in for availability
1 * 3 Items
Product Details & Documents

Borosilicate 9.1 glass.

  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in high contrast amber stain

 : DIN EN ISO 648

 : Dated batch identification and batch certificate.

Specifications
  • Kapacitet:
    2 ml
  • Længde:
    330 mm
  • Seq:
    02
  • Tolerance:
    0,010 ± ml