Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR® Bulb Pipettes, Two Marks, Class AS

1 ml, 0,007 ± ml, 300 mm
  612-4775
VWR® Bulb Pipettes, Two Marks, Class AS
  612-4775
VWR® Bulb Pipettes, Two Marks, Class AS
VWR® Bulb Pipettes, Two Marks, Class AS

1 ml, 0,007 ± ml, 300 mm

 
164,00 kr
   
1 * 3 Items
 

Borosilicate 9.1 glass.

  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in high contrast amber stain

Certifications: DIN EN ISO 648

Ordering information: Dated batch identification and batch certificate.

 
  • Kapacitet:
    1 ml
  • Længde:
    300 mm
  • Seq:
    01
  • Tolerance:
    0,007 ± ml