Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® Graduated Pipettes, Type 3, Class AS

VWR® Graduated Pipettes, Type 3, Class AS

25 ml, 0,1 ml, 0,15 ± ml
Katalog # 612-4773
Leverandør:   VWR Collection
VWR® Graduated Pipettes, Type 3, Class AS
Katalog # 612-4773
Leverandør:  VWR Collection
VWR® Graduated Pipettes, Type 3, Class AS
VWR® Graduated Pipettes, Type 3, Class AS

25 ml, 0,1 ml, 0,15 ± ml

Price
224,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents

Borosilicate 9.1 glass.

  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in amber stain
  • Cotton plug ending (cotton plugs not included)

Certifications: DIN 12696, ISO 835

Bestillingsoplysninger: Includes a lot certificate.

Specifikationer
  • Kapacitet:
    25 ml
  • Seq:
    07
  • Inddeling:
    0,1 ml
  • Tolerance:
    0,15 ± ml
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.