Push-in caps for tubes
LDPE, Caps, Ø×H; 25×10,5 mm, ufarvet
Catalog # SEMA7706
Supplier: SEMADENI
Jump to:
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifications
- Farve:ufarvet
- Beskrivelse:Caps, Ø×H; 25×10,5 mm
- Materiale:LDPE
- Seq:19