Orders LinkContactUsLinkComponent
Push-in caps for tubes
Push-in caps for tubes
LDPE, Caps, Ø 22 mm, ufarvet
Katalog # SEMA7702
Leverandør:   SEMADENI
Push-in caps for tubes
Katalog # SEMA7702
Leverandør:  SEMADENI
Push-in caps for tubes
Push-in caps for tubes
LDPE, Caps, Ø 22 mm, ufarvet
Price
404,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifikationer
  • Farve:
    ufarvet
  • Beskrivelse:
    Caps, Ø 22 mm
  • Materiale:
    LDPE
  • Seq:
    18