Orders LinkContactUsLinkComponent
Samplers, PowderThief
Samplers, PowderThief
Sampler body, 600 mm, 25,0 mm
Catalog # SAMP1030C-600
Supplier:   Sampling Systems
Samplers, PowderThief
Catalog # SAMP1030C-600
Supplier:  Sampling Systems
Samplers, PowderThief
Samplers, PowderThief
Sampler body, 600 mm, 25,0 mm
Price
5.310,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These PowderThief samplers are suitable for sampling free flowing powders and granules.
  • Made of high quaility 316 stainless steel
  • PTFE used for clutch mechanism
  • Single point sampler
A range of tips are available so that a single sampler can be used to accurately sample a range of different volumes.
Specifications
  • Ø:
    25,0 mm
  • Beskrivelse:
    Sampler body
  • Længde:
    600 mm
  • Seq:
    08
Accessories
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0495
    Nylon
    15 - 30 mm
    600 mm
    Log in for availability
    1 * 1 Items - 334,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SAMP8505A-01
    Standardprøvetagerbeholder, robust plastik, med fire glidelåse, 1.210 × 225 × 96 mm
    Log in for availability
    1 * 1 Items - 1.030,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0516
    Nylon
    7 - 14 mm
    600 mm
    Log in for availability
    1 * 1 Items - 334,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0496
    Nylon
    30 - 45 mm
    1100 mm
    Log in for availability
    1 * 1 Items - 418,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0515
    Nylon
    7 - 14 mm
    2100 mm
    Log in for availability
    1 * 1 Items - 585,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0498
    Nylon
    30 - 45 mm
    2100 mm
    Log in for availability
    1 * 1 Items - 585,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0497
    Nylon
    30 - 45 mm
    1600 mm
    Log in for availability
    1 * 1 Items - 501,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0499
    Nylon
    30 - 45 mm
    600 mm
    Log in for availability
    1 * 1 Items - 334,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0492
    Nylon
    15 - 30 mm
    1100 mm
    Log in for availability
    1 * 1 Items - 418,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0494
    Nylon
    15 - 30 mm
    2100 mm
    Log in for availability
    1 * 1 Items - 585,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0493
    Nylon
    15 - 30 mm
    1600 mm
    Log in for availability
    1 * 1 Items - 501,00 kr