Orders LinkContactUsLinkComponent
Accessories: Light and control
Accessories: Light and control
Knob motor, KM-01, to control the focus knob without touching the device, controlled using either a control box (included) or with DinoCapture 2.0 software, compatible with certain Dino-Lite models only
Katalog # IDCPKM-01
Leverandør:   Dino-Lite / IDCP B.V.
Accessories: Light and control
Katalog # IDCPKM-01
Leverandør:  Dino-Lite / IDCP B.V.
Accessories: Light and control
Accessories: Light and control
Knob motor, KM-01, to control the focus knob without touching the device, controlled using either a control box (included) or with DinoCapture 2.0 software, compatible with certain Dino-Lite models only
Price
3.940,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Knob motor, KM-01, to control the focus knob without touching the device, controlled using either a control box (included) or with DinoCapture 2.0 software, compatible with certain Dino-Lite models only