Vials for environmental analysis, WHEATON®
20,00 ml, 28×60 mm, klar, Yes, Open top, 24-400, PP, hvid, PTFE faced silicone
WHEAW224609
- Kasket inkluderet:Yes
- Hættemateriale:PP
- Cap størrelse:24-400
- Diameter×H:28×60 mm
- Septum:PTFE faced silicone
- Kapacitet:20,00 ml
- Farve:klar
- Seq:01
- Låg type:Open top
- Lågfarve:hvid