Orders LinkContactUsLinkComponent
Accessories for storage cabinets
3 shelves, 1 bottom collecting sump (V=21 l), stainless steel 1.4301
  MSGA30260
 :   asecos
Accessories for storage cabinets
  MSGA30260
 :  asecos
Accessories for storage cabinets
Accessories for storage cabinets
3 shelves, 1 bottom collecting sump (V=21 l), stainless steel 1.4301
3.310,00 kr
1 * 1 Each
  • Beskrivelse:
    3 shelves, 1 bottom collecting sump (V=21 l), stainless steel 1.4301
  • Seq:
    04