Accessories for storage cabinets
4 shelves, 1 perforated insert, 1 bottom collecting sump (V=21 l), stainless steel 1.4301
Katalog # MSGA30258
Leverandør: asecos
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:4 shelves, 1 perforated insert, 1 bottom collecting sump (V=21 l), stainless steel 1.4301
- Seq:14