Orders LinkContactUsLinkComponent
Accessories for storage cabinets
Accessories for storage cabinets
4 shelves, 1 perforated insert, 1 bottom collecting sump (V=22,5 l), sheet steel plastic laminated RAL 7035
Katalog # MSGA30250
Accessories for storage cabinets
Katalog # MSGA30250
Leverandør:  
Accessories for storage cabinets
Accessories for storage cabinets
4 shelves, 1 perforated insert, 1 bottom collecting sump (V=22,5 l), sheet steel plastic laminated RAL 7035
Discontinued
Product Details & Documents
Specifikationer
  • Beskrivelse:
    4 shelves, 1 perforated insert, 1 bottom collecting sump (V=22,5 l), sheet steel plastic laminated RAL 7035
  • Seq:
    15