Orders LinkContactUsLinkComponent
Bunded spill flooring
Bunded spill flooring, 80 l, wheel load 450 kg, 1000×1000×125 mm
  MSGA8812
 :   asecos
Bunded spill flooring
  MSGA8812
 :  asecos
Bunded spill flooring
Bunded spill flooring
Bunded spill flooring, 80 l, wheel load 450 kg, 1000×1000×125 mm
4.330,00 kr
1 * 1 Items
The bunded spill flooring made from galavanised steel is designed for covering a large storage area.
  • Removable, hot-dip galvanized grids
  • Optimal adaption on space demands
  • Approved for the safe storage of water pollutants and flammable liquids
Load capacity: 5 000 kg/m²
  • Beskrivelse:
    Bunded spill flooring, 80 l, wheel load 450 kg
  • Seq:
    03
  • B×D×H:
    1000×1000×125 mm
Accessories
  •  
    Beskrivelse
  • MSGA8819
    Joining element, 2470×45 mm
    1 * 1 Items - 520,00 kr
  •  
    Beskrivelse
  • MSGA8820
    Ramp, 450×1120 mm
    1 * 1 Items - 2.250,00 kr
  •  
    Beskrivelse
  • MSGA8821
    Ramp, 950×1120 mm
    1 * 1 Items - 4.060,00 kr
  •  
    Beskrivelse
  • MSGA2595
    Cross joiner, 130×130 mm
    1 * 1 Items - 436,00 kr
  •  
    Beskrivelse
  • MSGA8816
    Joining element, 470×45 mm
    1 * 1 Items - 257,00 kr
  •  
    Beskrivelse
  • MSGA8817
    Joining element, 970×45 mm
    1 * 1 Items - 348,00 kr
  •  
    Beskrivelse
  • MSGA8818
    Joining element, 1970×45 mm
    1 * 1 Items - 421,00 kr
  •  
    Beskrivelse
  • MSGA8822
    Ramp, 1950×1120 mm
    1 * 1 Items - 7.230,00 kr
  •  
    Beskrivelse
  • MSGA8823
    Ramp, 2450×1120 mm
    1 * 1 Items - 8.060,00 kr
  •  
    Beskrivelse
  • MSGA8824
    Distance fastener, 25×120 mm
    1 * 1 Items - 291,00 kr
  •  
    Beskrivelse
  • MSGA8825
    Edge fastener, 20×100 mm
    1 * 1 Items - 283,00 kr
  •  
    Beskrivelse
  • MSGA8826
    Corner ramp, 1120×1120 mm
    1 * 1 Items - 4.820,00 kr