Accessories for storage cabinets
3× shelf, load capacity: 100 kg, sheet steel galvanised; 1× bottom collecting sump, capacity: 33 L, sheet steel galvanised; weight: 20,5 kg
Katalog # 135-2915
Leverandør: asecos
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:3× shelf, load capacity: 100 kg, sheet steel galvanised; 1× bottom collecting sump, capacity: 33 L, sheet steel galvanised; weight: 20,5 kg
- Seq:01