Orders LinkContactUsLinkComponent
Accessories for storage cabinets
Accessories for storage cabinets
3× shelf, load capacity: 100 kg, sheet steel galvanised; 1× bottom collecting sump, capacity: 33 L, sheet steel galvanised; weight: 20,5 kg
Katalog # 135-2915
Leverandør:   asecos
Accessories for storage cabinets
Katalog # 135-2915
Leverandør:  asecos
Accessories for storage cabinets
Accessories for storage cabinets
3× shelf, load capacity: 100 kg, sheet steel galvanised; 1× bottom collecting sump, capacity: 33 L, sheet steel galvanised; weight: 20,5 kg
Price
1.350,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Specifikationer
  • Beskrivelse:
    3× shelf, load capacity: 100 kg, sheet steel galvanised; 1× bottom collecting sump, capacity: 33 L, sheet steel galvanised; weight: 20,5 kg
  • Seq:
    01