Mobile sumps for storage containers
Mobile sump for drums and small containers, galvanized, for flammable and water hazardous liquids, with 60 l sump capacity and without grid, Galvaniseret Stål, 635×810×980 mm
Katalog # MSGA23042
Leverandør: asecos
Gå til:
Product Details & Documents
Painted or galvanised steel.
- Easy to move by 2 swivel and 2 fixed wheels and push bar
For the storage of flammable and water hazardous liquids.
Specifikationer
- Beskrivelse:Mobile sump for drums and small containers, galvanized, for flammable and water hazardous liquids, with 60 l sump capacity and without grid
- Materiale:Galvaniseret Stål
- Seq:02
- B×D×H:635×810×980 mm