Orders LinkContactUsLinkComponent
Mobile sumps for storage containers
Mobile sump for drums and small containers, painted, for flammable and water hazardous liquids, with 60 l sump capacity and without grid, 635×810×980 mm
Catalog # MSGA23041
Supplier:   asecos
Mobile sumps for storage containers
Catalog # MSGA23041
Supplier:  asecos
Mobile sumps for storage containers
Mobile sump for drums and small containers, painted, for flammable and water hazardous liquids, with 60 l sump capacity and without grid, 635×810×980 mm
Price
3.070,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Painted or galvanised steel.
  • Easy to move by 2 swivel and 2 fixed wheels and push bar
For the storage of flammable and water hazardous liquids.
Specifications
  • Beskrivelse:
    Mobile sump for drums and small containers, painted, for flammable and water hazardous liquids, with 60 l sump capacity and without grid
  • Seq:
    01
  • B×D×H:
    635×810×980 mm