Dialysis flasks, Slide-A-Lyzer™
These dialysis flasks facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 ml.
- Easy to use
- Color-coded for easy identification
- No leakage, sample remains intact with no contamination
- High surface area to volume ratio maximizes diffusion rate
Devices are constructed from two sheets of low protein-binding, regenerated cellulose membranes to ensure maximum sample recovery and purity and contain up to 85% less plastic per volume compared to other devices. Slide-A-Lyzer dialysis flasks are compatible with most laboratory buffers and can accomplish low-molecular weight contaminant removal, buffer exchange and desalting in 8 hours to overnight.
- MWCO:20 kD
- Farve:lilla
- Beskrivelse:Slide-A-Lyzer dialysis flasks
- Seq:04
- Prøvevolumen:150 - 250 ml
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



