Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dialysis flasks, Slide-A-Lyzer™

Dialysis flasks, Slide-A-Lyzer™

Slide-A-Lyzer dialysis flasks, 2 kD, blå, 150 - 250 ml
Katalog # PIER87760
Leverandør:   Thermo Fisher Scientific
Dialysis flasks, Slide-A-Lyzer™
Katalog # PIER87760
Dialysis flasks, Slide-A-Lyzer™
Dialysis flasks, Slide-A-Lyzer™

Slide-A-Lyzer dialysis flasks, 2 kD, blå, 150 - 250 ml

Price
3.480,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents

These dialysis flasks facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 ml.

  • Easy to use
  • Color-coded for easy identification
  • No leakage, sample remains intact with no contamination
  • High surface area to volume ratio maximizes diffusion rate

Devices are constructed from two sheets of low protein-binding, regenerated cellulose membranes to ensure maximum sample recovery and purity and contain up to 85% less plastic per volume compared to other devices. Slide-A-Lyzer dialysis flasks are compatible with most laboratory buffers and can accomplish low-molecular weight contaminant removal, buffer exchange and desalting in 8 hours to overnight.

Specifikationer
  • MWCO:
    2 kD
  • Farve:
    blå
  • Beskrivelse:
    Slide-A-Lyzer dialysis flasks
  • Seq:
    01
  • Prøvevolumen:
    150 - 250 ml
Accessories

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Flotation disks for Slide-A-Lyzer™ dialysis flasks, LDPE
    Flotation disks for Slide-A-Lyzer™ dialysis flasks, LDPE
    PIER87759
    Log ind for tilgængelighed
    1 * 6 Items - 277,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.