Orders LinkContactUsLinkComponent
Safety goggles, 602
Safety goggles, 602
Anti-UV, uncoated, Clear PC, klar
Katalog # UNVT602.01.00.01
Leverandør:   UNIVET
Safety goggles, 602
Katalog # UNVT602.01.00.01
Leverandør:  UNIVET
Safety goggles, 602
Safety goggles, 602
Anti-UV, uncoated, Clear PC, klar
Price
333,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
The 602 goggles have an essential frame which is ideal for each type of fit.
  • Adjustable elastic fabric headband
  • Polycarbonate lens
  • Indirect ventilation system
  • OTG (worn over prescription glasses)
These goggles are used for mechanical works in good visibility conditions.
Wearing these goggles for prolonged period of time will not create distortion or fatigue.
Weight: 73 g
Specifikationer
  • Model:
    Anti-UV, uncoated
  • Farve:
    klar
  • Seq:
    02
  • Linser:
    Clear PC