Swabs in tubes
Swabs in tube with mini-tip, Plast / polyester, +, 150 mm
Katalog # GREI421008
Gå til:
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
Raynon Swab i Peeling Pose m/Plast Greb 1 * 1.000 Items Katalog #710-0186
Produktet 710-0186 ligner GREI421008, men producenten og specifikationerne kan variere.
Product Details & Documents
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
- Sterilised by irradiation
- Tubes are made of polystyrene
- Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Steril:+
- Beskrivelse:Swabs in tube with mini-tip
- Længde:150 mm
- Seq:03
- Skaft/hoved:Plast / polyester