Orders LinkContactUsLinkComponent
Swabs in tubes
Swabs in tubes
Swabs in tube with mini-tip, Plast / polyester, +, 150 mm
Katalog # GREI421008
Swabs in tubes
Katalog # GREI421008
Leverandør:  
Swabs in tubes
Swabs in tube with mini-tip, Plast / polyester, +, 150 mm
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
Raynon Swab i Peeling Pose m/Plast Greb 1 * 1.000 Items Katalog #710-0186
Produktet 710-0186 ligner GREI421008, men producenten og specifikationerne kan variere.
Product Details & Documents
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
  • Sterilised by irradiation
  • Tubes are made of polystyrene
  • Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Documents
Specifikationer
  • Steril:
    +
  • Beskrivelse:
    Swabs in tube with mini-tip
  • Længde:
    150 mm
  • Seq:
    03
  • Skaft/hoved:
    Plast / polyester