Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Culture flasks, Erlenmeyer shape, BIOGEN

50 ml, 85 mm, 51 mm, 38 mm
  WITG5.560.050
Culture flasks, Erlenmeyer shape, BIOGEN
  WITG5.560.050
Culture flasks, Erlenmeyer shape, BIOGEN
Culture flasks, Erlenmeyer shape, BIOGEN

50 ml, 85 mm, 51 mm, 38 mm

 
2.570,00 kr
   
1 * 10 Items
 

Culture flask, made of borosilicate glass, Erlenmeyer shape, with straight neck for metal caps.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
 
  • Udv.Ø:
    51 mm
  • Kapacitet:
    50 ml
  • Højde:
    85 mm
  • Seq:
    01
  • Hals Ø udv.:
    38 mm