Push-in caps for tubes
LDPE, Caps, Ø×H; 12,5×7,5 mm, ufarvet
SEMA7701
: SEMADENI
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
- Farve:ufarvet
- Beskrivelse:Caps, Ø×H; 12,5×7,5 mm
- Materiale:LDPE
- Seq:10