Micro cover glasses
Cover glass 2450-1.5
Katalog # 630-2097
Leverandør: LEICA HISTOLOGY
Gå til:
Product Details & Documents
These micro cover glasses feature a fine pre-cleaned borosilicate glass.
- Uniform thinness and flatness
- Non corrosive and anti-fogging glass in a lint-free, shock absorbing lining
- Strict specification control during the cutting, cleaning, weighing and packaging procedures
Emballering: Includes dessicant.
Specifikationer
- Beskrivelse:Cover glass 2450-1.5