Orders LinkContactUsLinkComponent
Micro cover glasses
Micro cover glasses
Cover glass 4570-1.5
Katalog # 630-2098
Leverandør:   LEICA HISTOLOGY
Micro cover glasses
Katalog # 630-2098
Leverandør:  LEICA HISTOLOGY
Micro cover glasses
Cover glass 4570-1.5
Price
48,30 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
These micro cover glasses feature a fine pre-cleaned borosilicate glass.
  • Uniform thinness and flatness
  • Non corrosive and anti-fogging glass in a lint-free, shock absorbing lining
  • Strict specification control during the cutting, cleaning, weighing and packaging procedures
Emballering: Includes dessicant.
Specifikationer
  • Beskrivelse:
    Cover glass 4570-1.5