Hydrogenperoxid 30%, Suprapur® for trace analysis, Supelco®
Plastic bottle, 500 mL
Leverandør: Merck
Hydrogenperoxid 30%, Suprapur® for trace analysis, Supelco®
Katalog # 1.07298.0500
Leverandør: Merck
CAS nummer: 7722-84-1
Gå til:
Product Details & Documents
- CAS nummer:7722-84-1
- Formel:H₂O₂
- MDL nummer:MFCD00011333
- Molecular Weight:34.01 g/mol
- Kogende Pt:∼107 °C (1013 hPa)
- Smeltende Pt:< 0 °C
- Tæthed:1,11 g/cm³ (20 °C)
- ADR:5.1 (8),II
- :M-Clarity™ quality level = MQ100
- :Density: 1.11 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-765-0
- FN:2014
Documents
Certifikater
Søg efter partinummerMERC1.07298.0250_TDS_EN MERC1.07298.0500_TDS_EN MERC1.07298.1000_TDS_EN ProductPage_107298
Produktspecifikation
Merck Pharma Segment Brochure Merck Ultrapur and Suprapur® High Purity Acids and Bases
Produktlitteratur
msds_107298
SDS
Specifikationer
Specifikationstestresultater
- Solubility:(20 °C) soluble
- Melting Point:-26 °C
- Boiling Point:107 °C (1013 hPa)
- Vapor Pressure:18 hPa (20 °C)
- Density:1.11 g/cm3 (20 °C)
- pH:2 - 4 (H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



