Touch screen gloves, GL16
L/XL, sort/grå
Katalog # 112-2950
Leverandør: Portwest
Gå til:
Product Details & Documents
These knitted gloves are made of 100% acrylic fibres. The black acrylic parts are hardwearing while the grey acrylic with metallic component allows for precise movements on a touch screen device.
- High dexterity
- Grey acrylic fabric on thumb and two fingers
Specifikationer
- Handske størrelse:L/XL
- Farve:sort/grå
- Seq:02
- Cat. No.:112-2950
- Ambidextrous/Hånd-specifik:Hånd-specifik
- Model:GL16
- Manchet:Strikket
- Engangs/Genbrugelig:Genanvendelig