Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Winter overalls, Essential

Winter overalls, Essential

grå/sort, XL, Regular
Katalog # KWIN100824896XL
Leverandør:   FRISTADS KANSAS
Winter overalls, Essential
Katalog # KWIN100824896XL
Leverandør:  FRISTADS KANSAS
Winter overalls, Essential
Winter overalls, Essential

grå/sort, XL, Regular

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Protective, bi-coloured overalls made of 100% polyester. With quilted lining in body.

  • Metal zip closure, collar with fur fibre lining and detachable, adjustable hood
  • Two chest and two front pockets with flap, back pocket with flap, two inside pockets
  • Mobile phone pocket, pen pocket, folding-ruler pocket
  • Leg zipper, leg ends adjustable by press-studs
  • Reflective piping on the yoke

Weight: 200 g/m²

Documents

Specifikationer
  • Tøj størrelse:
    XL
  • Beklædningsgenstand:
    Regular
  • Farve:
    grå/sort
  • Seq:
    17
  • Køn:
    Unisex
  • Cat. No.:
    KWIN100824896XL
  • Sleeve or leg length:
    Long
  • Risk area:
    Cold
  • Vægt:
    200 g/m²
  • Lukkestørrelse:
    Med lynlås
  • Stof:
    Polyester
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.