Sweatshirt, Acode® Basecamp 1734
rød, L
Katalog # KWIN100225331L
Leverandør: FRISTADS KANSAS
Gå til:
Product Details & Documents
Sweatshirt made of 80% cotton and 20% polyester, brushed inside.
- Round neck
- Neck, collar, cuffs and hem in rib with elastane
Weight: 280 g/m²
Documents
KWIN100225740S_TDS_EN
Datasheet
Specifikationer
- Tøj størrelse:L
- Farve:rød
- Seq:29
- Køn:For mænd
- Cat. No.:KWIN100225331L
- Sleeve or leg length:Long
- Risk area:Cold
- Vægt:280 g/m²
- Model:1734
- Outfit applikationer:Industrial
- Stof:80% cotton/20% polyester