Orders LinkContactUsLinkComponent
Safety cones, blank
Cone, orange, height: 89 cm
Catalog # 116-0335
Supplier:   Brady
Safety cones, blank
Catalog # 116-0335
Supplier:  Brady
Safety cones, blank
Safety cones, blank
Cone, orange, height: 89 cm
Price
827,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Attachable signs warn against particular hazards and provide good safety lighting for dark areas. Barrier chains can be attached to the cones.
  • Visible from all four directions
  • Easy to stack for storage
The materials used, which are resistant to impact and to weather conditions, are suitable for both indoor and outdoor use.
Specifications
  • Beskrivelse:
    Cone, orange, height: 89 cm