Chains, Bradylink™
Chain, black/yellow, PE, 50×25000 mm
Katalog # 116-0345
Leverandør: Brady
Gå til:
Product Details & Documents
Bradylink™ chains are suitable for use with the posts and connection hooks.
- Various colours available
Specifikationer
- Beskrivelse:Chain, black/yellow
- Dimensioner:50×25000 mm
- Materiale:PE