Work jackets, Icon
grå/sort, M
Catalog # KWIN100811896M
Supplier: FRISTADS KANSAS
Jump to:
Product Details & Documents
Bi-coloured workers jacket made of 100% cotton.
- Contrast stitching
- Button front with storm flap
- Angle-cut sleeves, adjustable cuffs and waistband
- Broad back with yoke, reflector wings in side seams
- Two chest pockets with pouch and flap, mobile phone pocket with zip and storm flap, inside pocket
- Diagonal cut on front with integrated side pockets with internal pouch
Weight: 375 g/m²
Documents
Specifications
- Tøj størrelse:M
- Farve:grå/sort
- Seq:10
- Køn:Unisex
- Cat. No.:KWIN100811896M
- Sleeve or leg length:Long
- Vægt:375 g/m²
- Outfit applikationer:Industrial
- Lukkestørrelse:Med knapper
- Stof:100% cotton