Jacket, 762 P
mørkeblå, XL
Katalog # KWIN100461540XL
Leverandør: FRISTADS KANSAS
Gå til:
Product Details & Documents
These reversible jackets are made of 100% polyester fur fibre.
- Zip to top of collar
- Thumb grip
- Lengthened back
Weight: 320 g/m²
Documents
Specifikationer
- Tøj størrelse:XL
- Farve:mørkeblå
- Køn:Unisex
- Cat. No.:KWIN100461540XL
- Sleeve or leg length:Long
- Risk area:Cold
- Vægt:320 g/m²
- Model:Crafts 762 P
- Outfit applikationer:General Purpose
- Lukkestørrelse:Med lynlås
- Stof:100% polyester fur