Orders LinkContactUsLinkComponent
First aid cabinets
First aid cabinets
Mini first aid panel, 250×50×400 mm
Katalog # 118-0254
First aid cabinets
Katalog # 118-0254
First aid cabinets
First aid cabinets
Mini first aid panel, 250×50×400 mm
Price
787,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
First aid cabinet with double door (118-0261)
This metal cabinet has two doors that divide the cabinet into two sections. The outer section is organised as a first aid panel with easily accessible products. A plexiglass door protects the products from dust and moisture. The inner section of the cabinet consists of a lockable space for a substantial stock of refill products.
Mini first aid cabinet with panel (118-0262)
This small metal cabinet comes with a mini first aid panel fixed on the door. Refills are stored in the lockable cabinet.
Mini first aid panel (118-0254)
This green, hard plastic panel can either be be put on the wall or attached to the mini first aid cabinet. The panel contains of a variety of first aid products and a first aid instruction.
Colour: Green
Specifikationer
  • Beskrivelse:
    Mini first aid panel, 250×50×400 mm