Orders LinkContactUsLinkComponent
Objectives for PSM-1000 microscopes
Objectives for PSM-1000 microscopes
Near Infrared Ray (NIR) objective (WD = 20,5 mm), 20×/0,4
Catalog # MOTI1101001703531
Supplier:   MOTIC
Objectives for PSM-1000 microscopes
Catalog # MOTI1101001703531
Supplier:  MOTIC
Objectives for PSM-1000 microscopes
Near Infrared Ray (NIR) objective (WD = 20,5 mm), 20×/0,4
Price
15.590,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Forstørrelse:
    20×/0,4
  • Beskrivelse:
    Near Infrared Ray (NIR) objective (WD = 20,5 mm)
  • Seq:
    111