Orders LinkContactUsLinkComponent
Disposable earplugs, Contours®
Earplugs, without cord, in dispenser MoldexStation, gul/grøn, M
Katalog # 111-1169
Leverandør:   Moldex
Disposable earplugs, Contours®
Katalog # 111-1169
Leverandør:  Moldex
Disposable earplugs, Contours®
Disposable earplugs, Contours®
Earplugs, without cord, in dispenser MoldexStation, gul/grøn, M
Price
1.140,00 kr
Log ind for tilgængelighed
1 * 500 Par
Product Details & Documents
These extremely comfortable earplugs made of soft polyurethane foam are available in two different sizes.
  • Anatomical shape easily adapts to the ear canal: Less pressure and more comfort
  • Large base helps provide proper insertion, fit, and removal
  • Bright colour for improved visibility
SNR: 35 dB
 : EN 352-2:2002
 : Supplied in a box or in the hygienic dispenser system MoldexStation.
 : In pairs.
Specifikationer
  • Farve:
    gul/grøn
  • Beskrivelse:
    Earplugs, without cord, in dispenser MoldexStation
  • Størrelse:
    M
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Wall-mounting bracket for MoldexStation earplug dispenser
    Wall-mounting bracket for MoldexStation earplug dispenser
    111-0724
    Log ind for tilgængelighed
    1 * 1 Items - 77,20 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • CONTOURS® refill pack
    CONTOURS® refill pack
    MOLD746001
    Log ind for tilgængelighed
    1 * 500 Par - 1.080,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • CONTOURS® small refill pack
    CONTOURS® small refill pack
    MOLD746301
    Log ind for tilgængelighed
    1 * 500 Par - 1.080,00 kr