Safety spectacles, Virtua™ AP
Anti-UV, anti-scratch, transparent, Clear PC
Katalog # 111-2281
Leverandør: 3M
Gå til:
Product Details & Documents
Lightweight, stylish designed safety glasses Virtua™ AP protective eyewear features a streamline wrap-around design for excellent coverage and good field of vision.
- Integrated brow bar for added protection
- Lightweight for maximum comfort
- Strong polycarbonate lenses featuring anti-scratch coating for improved vision and extended durability
Weight: 26 g
: EN 166:2001
Specifikationer
- Model:Anti-UV, anti-scratch
- Farve:transparent
- Seq:01
- Linser:Clear PC