Orders LinkContactUsLinkComponent
Safety spectacles, Virtua™ AP
Safety spectacles, Virtua™ AP
Anti-UV, anti-scratch, transparent, Clear PC
Katalog # 111-2281
Leverandør:   3M
Safety spectacles, Virtua™ AP
Katalog # 111-2281
Leverandør:  3M
Safety spectacles, Virtua™ AP
Safety spectacles, Virtua™ AP
Anti-UV, anti-scratch, transparent, Clear PC
Price
34,20 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Lightweight, stylish designed safety glasses Virtua™ AP protective eyewear features a streamline wrap-around design for excellent coverage and good field of vision.
  • Integrated brow bar for added protection
  • Lightweight for maximum comfort
  • Strong polycarbonate lenses featuring anti-scratch coating for improved vision and extended durability
Weight: 26 g
 : EN 166:2001
Specifikationer
  • Model:
    Anti-UV, anti-scratch
  • Farve:
    transparent
  • Seq:
    01
  • Linser:
    Clear PC