Inserts
E 129-06 insert for infusion bottles, 20 fields, square measure 84×84 mm, PG 8535/36, 445×445×113 mm
Katalog # MIEL3808550
Leverandør: MIELE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:E 129-06 insert for infusion bottles, 20 fields, square measure 84×84 mm
- Seq:42
- B×D×H:445×445×113 mm
- Til:PG 8535/36