Inserts
E 126-06 insert for infusion bottles, 48 fields, square measure 45×45 mm, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 445×445×83 mm
Catalog # MIEL3808520
Supplier: MIELE
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:E 126-06 insert for infusion bottles, 48 fields, square measure 45×45 mm
- Seq:40
- B×D×H:445×445×83 mm
- Til:PG 8535/36, PG 8582, PG 8582 CD, PG 8592