Inserts
E 125-06 insert for infusion bottles, 9 fields, square measure 125×125 mm, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 460×460×224 mm
Katalog # MIEL3808510
Leverandør: MIELE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:E 125-06 insert for infusion bottles, 9 fields, square measure 125×125 mm
- Seq:39
- B×D×H:460×460×224 mm
- Til:PG 8535/36, PG 8582, PG 8582 CD, PG 8592