Inserts
E 124-06 insert for infusion bottles, 16 fields, square measure 100x100 mm, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 460×460×148 mm
Katalog # MIEL3808500
Leverandør: MIELE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:E 124-06 insert for infusion bottles, 16 fields, square measure 100x100 mm
- Seq:38
- B×D×H:460×460×148 mm
- Til:PG 8535/36, PG 8582, PG 8582 CD, PG 8592