Accessories
Upgrade AD (demineralised water), All Miele glassware units
Katalog # 141-0504
Gå til:
Discontinued
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifikationer
- Beskrivelse:Upgrade AD (demineralised water)
- Seq:48
- Til:All Miele glassware units