Orders LinkContactUsLinkComponent
Inserts
Inserts
E 402 Insert suitable for 44 watch glasses Ø 80 - 125 mm, equipped with 23 bars spaced 15 mm apart, W×D×H: 600×805×1003 mm
Katalog # 141-0387
Leverandør:   MIELE
Inserts
Katalog # 141-0387
Leverandør:  MIELE
Inserts
Inserts
E 402 Insert suitable for 44 watch glasses Ø 80 - 125 mm, equipped with 23 bars spaced 15 mm apart, W×D×H: 600×805×1003 mm
Price
3.380,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    E 402 Insert suitable for 44 watch glasses Ø 80 - 125 mm, equipped with 23 bars spaced 15 mm apart, W×D×H: 600×805×1003 mm
  • Seq:
    51