Inserts
E 144 Insert suitable for 18 glass beakers, equipped with 18×3 stay bars, W×D×H: 200x445x131 mm
Katalog # 141-0299
Leverandør: MIELE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:E 144 Insert suitable for 18 glass beakers, equipped with 18×3 stay bars, W×D×H: 200x445x131 mm
- Seq:43